Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ins-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065510.7 | NP_497911.1 | 1 - 2 | 211 | 74 | |
GenBank mRNA | AB009658.1 | - | 1 - 2 | 200 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |