Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpg-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025988.7 | NP_001021159.1 | 1 - 2 | 339 | 83 | |
NM_001025989.5 | NP_001021160.1 | 1 - 2 | 246 | 83 | ||
GenBank mRNA | DQ340623.1 | - | 1 - 2 | 246 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |