Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mab-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065539.6 | NP_497940.2 | 3 - 4 | 519 | 99 | |
GenBank mRNA | U19861.2 | - | 3 - 4 | 514 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |