Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rps-0 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065577.8 | NP_497978.1 | 1 - 2 | 139 | 69 |
Gene Symbol: B0393.8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381844.1 | NP_001366937.1 | 1 - 2 | 145 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |