Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ced-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026031.4 | NP_001021202.1 | 1 - 2 | 444 | 130 | |
NM_001026032.3 | NP_001021203.1 | 1 - 2 | 444 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |