Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ced-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026031.4 | NP_001021202.1 | 7 - 8 | 1422 | 61 | |
NM_001026032.3 | NP_001021203.1 | 7 - 8 | 1488 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |