Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ula-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065636.5 | NP_498037.2 | 5 - 6 | 835 | 98 |
Gene Symbol: mmab-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001384085.1 | 9 - 10 | 1866 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |