Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: set-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065638.6 | NP_498039.1 | 10 - 11 | 2552 | 66 | |
NM_065639.6 | NP_498040.1 | 10 - 11 | 2616 | 66 | ||
NM_065640.6 | NP_498041.1 | 2 - 3 | 344 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |