Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: set-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065638.6 | NP_498039.1 | 5 - 6 | 859 | 121 | |
NM_065639.6 | NP_498040.1 | 5 - 6 | 932 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |