Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gop-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065717.6 | NP_498118.1 | 4 - 5 | 929 | 82 | |
GenBank mRNA | AY052770.1 | - | 4 - 5 | 929 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |