Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqe-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047389.4 | NP_001040854.1 | 2 - 3 | 160 | 62 | |
NM_001382869.1 | 2 - 3 | 161 | 62 | |||
NM_065734.8 | NP_498135.2 | 2 - 3 | 167 | 62 | ||
NM_065735.6 | NP_498136.2 | 2 - 3 | 161 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |