Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: paa-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065761.5 | NP_498162.2 | 2 - 3 | 1380 | 55 | |
GenBank mRNA | AB110419.1 | - | 2 - 3 | 1369 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |