Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025977.6 | NP_001021148.1 | 3 - 4 | 400 | 76 | |
NM_001025978.3 | NP_001021149.1 | 3 - 4 | 397 | 76 | ||
NM_001322590.3 | NP_001309588.1 | 4 | 407 | 76 | ||
NM_065810.7 | NP_498211.1 | 3 - 4 | 397 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |