Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: par-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027436.3 | NP_001022607.1 | 7 - 8 | 1648 | 65 | |
NM_001306735.2 | NP_001293664.1 | 3 - 4 | 1330 | 65 | ||
NM_001306736.3 | NP_001293665.1 | 3 - 4 | 1330 | 65 | ||
NM_001306739.3 | NP_001293668.1 | 7 - 8 | 1648 | 65 | ||
NM_001306740.3 | NP_001293669.1 | 7 - 8 | 1648 | 65 | ||
NM_001306741.3 | NP_001293670.1 | 7 - 8 | 1648 | 65 | ||
NM_001372593.2 | NP_001360234.1 | 3 - 4 | 1330 | 65 | ||
NM_001372595.2 | NP_001360237.1 | 3 - 4 | 1330 | 65 | ||
NM_001379719.1 | NP_001368259.1 | 6 - 7 | 1567 | 65 | ||
NM_001379720.2 | NP_001367341.1 | 3 - 4 | 1459 | 65 | ||
NM_001379721.1 | NP_001368258.1 | 3 - 4 | 1330 | 65 | ||
NM_001379722.1 | NP_001368260.1 | 3 - 4 | 1330 | 65 | ||
NM_001382875.1 | 7 - 8 | 1644 | 65 | |||
GenBank mRNA | U25032.1 | - | 7 - 8 | 1647 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |