Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sma-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047399.4 | NP_001040864.1 | 5 - 6 | 710 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |