Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eng-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026067.5 | NP_001021238.1 | 2 - 3 | 299 | 120 | |
NM_001372601.3 | NP_001359901.1 | 2 - 3 | 277 | 120 | ||
NM_065866.6 | NP_498267.3 | 2 - 3 | 301 | 120 | ||
GenBank mRNA | AB079783.1 | - | 2 - 3 | 299 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |