Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eng-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026067.5 | NP_001021238.1 | 4 - 5 | 883 | 90 | |
NM_001372601.3 | NP_001359901.1 | 4 - 5 | 861 | 90 | ||
GenBank mRNA | AB079783.1 | - | 4 - 5 | 883 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |