Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npp-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379737.1 | NP_001367842.1 | 9 - 10 | 4892 | 71 | |
NM_065909.5 | NP_498310.3 | 9 - 10 | 4893 | 71 | ||
GenBank mRNA | GU174496.1 | - | 9 - 10 | 4882 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |