Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nfm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027400.4 | NP_001022571.1 | 6 - 7 | 1231 | 127 | |
NM_065934.5 | NP_498335.1 | 6 - 7 | 1657 | 127 | ||
NM_065935.5 | NP_498336.1 | 6 - 7 | 1621 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |