Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379767.1 | NP_001367477.1 | 5 - 6 | 602 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |