Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001368430.2 | NP_001355512.1 | 7 - 8 | 1153 | 65 | |
NM_001368431.3 | NP_001355513.1 | 7 - 8 | 1111 | 65 | ||
NM_001368432.3 | NP_001355514.1 | 6 | 1018 | 65 | ||
NM_001368433.3 | NP_001355515.1 | 6 | 1018 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |