Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdfr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025999.4 | NP_001021170.1 | 6 - 7 | 918 | 97 | |
NM_001026000.4 | NP_001021171.1 | 6 - 7 | 918 | 97 | ||
NM_001306692.3 | NP_001293621.1 | 4 - 5 | 281 | 97 | ||
NM_001372610.2 | NP_001360187.1 | 4 - 5 | 275 | 97 | ||
NM_001382898.1 | 4 - 5 | 275 | 97 | |||
GenBank mRNA | AY314776.1 | - | 6 - 7 | 918 | 97 | |
AY314777.1 | - | 6 - 7 | 918 | 97 | ||
AY314778.1 | - | 6 - 7 | 918 | 97 | ||
EF141316.1 | - | 3 - 4 | 196 | 97 | ||
EF141317.1 | - | 3 - 4 | 196 | 97 | ||
EF141318.1 | - | 3 - 4 | 196 | 97 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |