Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lim-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066083.7 | NP_498484.1 | 11 - 12 | 2179 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |