Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sma-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066092.4 | NP_498493.1 | 5 - 6 | 462 | 66 | |
GenBank mRNA | U34902.1 | - | 5 - 6 | 463 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |