Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kbp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306710.2 | NP_001293639.1 | 3 - 4 | 378 | 95 |
Gene Symbol: K04C2.8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027489.5 | NP_001022660.2 | 3 - 4 | 377 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |