Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpn-6.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027450.4 | NP_001022621.1 | 4 - 5 | 725 | 107 | |
NM_001027451.5 | NP_001022622.1 | 5 - 6 | 706 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |