Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpn-6.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027450.4 | NP_001022621.1 | 6 - 7 | 1359 | 149 | |
NM_001027451.5 | NP_001022622.1 | 7 - 8 | 1340 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |