Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001368434.4 | NP_001355511.1 | 12 | 1286 | 91 | |
NM_066131.8 | NP_498532.4 | 11 - 12 | 1255 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons |