Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpg-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066150.13 | NP_498551.3 | 2 - 3 | 121 | 64 | |
GenBank mRNA | DQ340624.1 | - | 1 - 2 | 69 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |