Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066153.7 | NP_498554.1 | 1 - 2 | 118 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |