Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025942.4 | NP_001021113.1 | 7 - 8 | 794 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |