Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clk-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382917.1 | 11 - 12 | 2177 | 93 | ||
GenBank mRNA | AF400665.1 | - | 3 - 4 | 507 | 93 | |
AJ320257.1 | - | 3 - 4 | 501 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |