Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lpd-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066260.6 | NP_498661.1 | 3 - 4 | 352 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |