Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cup-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066309.6 | NP_498710.2 | 4 - 5 | 740 | 98 | |
GenBank mRNA | AY611497.1 | - | 4 - 5 | 720 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |