Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cup-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066309.6 | NP_498710.2 | 6 - 7 | 1279 | 163 | |
GenBank mRNA | AY611497.1 | - | 6 - 7 | 1259 | 163 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |