Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dcr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066360.4 | NP_498761.2 | 19 - 20 | 4231 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |