Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047386.5 | NP_001040851.1 | 6 - 7 | 1131 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |