Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066497.5 | NP_498898.2 | 3 - 4 | 388 | 61 | |
GenBank mRNA | AY647457.1 | - | 3 - 4 | 388 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |