Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdhk-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066527.6 | NP_498928.1 | 5 - 6 | 745 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |