Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sma-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382944.1 | 5 - 6 | 474 | 78 | ||
GenBank mRNA | U10327.1 | - | 4 - 5 | 283 | 78 | |
U34778.1 | - | 3 - 4 | 239 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |