Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F54F2.14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379849.1 | NP_001368592.1 | 15 - 16 | 3830 | 81 |
Gene Symbol: zfp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382947.1 | 15 - 16 | 3927 | 81 | ||
NM_001382948.1 | 14 - 15 | 4367 | 81 | |||
NR_136383.1 | 14 - 15 | 3072 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |