Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F54F2.14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379849.1 | NP_001368592.1 | 9 - 10 | 1759 | 95 |
Gene Symbol: zfp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382947.1 | 9 - 10 | 1856 | 95 | ||
NM_001382948.1 | 8 - 9 | 2303 | 95 | |||
NR_136383.1 | 8 - 9 | 1001 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |