Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027844.4 | NP_001023015.1 | 6 - 7 | 1496 | 98 | |
NM_001027845.5 | NP_001023016.1 | 6 - 7 | 1493 | 98 | ||
GenBank mRNA | AF269693.1 | - | 6 - 7 | 1496 | 98 | |
AF269694.1 | - | 6 - 7 | 1490 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |