Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pde-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027534.6 | NP_001022705.2 | 7 - 8 | 1187 | 99 | |
NM_001027535.6 | NP_001022706.2 | 6 - 7 | 1082 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |