Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpn-6.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268043.2 | NP_001254972.1 | 3 - 4 | 1008 | 75 | |
NM_001268044.3 | NP_001254973.1 | 1 - 2 | 373 | 75 |
Gene Symbol: F59B2.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379854.1 | NP_001366671.1 | 3 - 4 | 1020 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |