Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zif-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066591.6 | NP_498992.2 | 1 - 2 | 83 | 126 | |
GenBank mRNA | AY547284.1 | - | 1 - 2 | 73 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |