Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eat-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306696.3 | NP_001293625.1 | 3 - 4 | 381 | 65 | |
NM_066622.6 | NP_499023.3 | 4 - 5 | 473 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |