Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eat-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306696.3 | NP_001293625.1 | 7 - 8 | 871 | 130 | |
NM_066622.6 | NP_499023.3 | 8 - 9 | 963 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |