Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322726.2 | NP_001309582.1 | 1 - 2 | 152 | 65 | |
NM_066627.6 | NP_499028.1 | 1 - 2 | 152 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |