Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dat-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066642.6 | NP_499043.1 | 10 - 11 | 1541 | 84 | |
GenBank mRNA | AF079899.1 | - | 10 - 11 | 1549 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |